Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GlyT1/SLC6A9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GlyT1/SLC6A9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GlyT1/SLC6A9 Polyclonal specifically detects GlyT1/SLC6A9 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GlyT1/SLC6A9 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
6536 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
DKFZp547A1118, GlyT1, GlyT-1, GLYT1glyT-1, sodium- and chloride-dependent glycine transporter 1, solute carrier family 6 (neurotransmitter transporter, glycine), member 9, Solute carrier family 6 member 9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human GlyT1/SLC6A9 (NP_964012.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title