Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GlyT2 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577457
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes a sodium- and chloride-dependent glycine neurotransmitter transporter. This integral membrane glycoprotein is responsible for the clearance of extracellular glycine during glycine-mediated neurotransmission.
Specifications
GlyT2 (extracellular) | |
Polyclonal | |
Unconjugated | |
Slc6a5 | |
SLC6A5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
104245, 171148 | |
-20°C | |
Lyophilized |
Immunofluorescence, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P58295, Q761V0 | |
Slc6a5 | |
Peptide CVIGDHPKIQIKNS, corresponding to amino acid residues 333-346 of rat Glycine Transporter 2 | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction