Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSF Antibody (83308), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5181A
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Rat samples. It is validated for Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
GM-CSF | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
83308 | |
Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
E. coli-derived recombinant rat GM-CSF, Ala1-Lys127, Accession # P48750 | |
0.5 mL | |
Biologically Active Proteins, Cytokine Research, Immunology | |
1437 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction