Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSF Antibody (OTI8G5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270462V
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GM-CSF | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CSF2 | |
Recombinant protein fragment corresponding to amino acids 18-144 of human GM-CSF (NP_000749) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI8G5 | |
Western Blot, Immunohistochemistry | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cytokine Research, Immunology | |
1437.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction