Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSF Antibody Pair (HRP), Novus Biologicals™

Solid phase sandwich ELISA for the quantitative determination of Human GM-CSF.
$854.00
Specifications
Antigen | GM-CSF |
---|---|
Dilution | Sandwich ELISA |
Classification | Monoclonal |
Conjugate | HRP |
Host Species | Mouse |
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
GM-CSF | |
Monoclonal | |
Mouse | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
CSF2 | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human GM-CSF. |
Sandwich ELISA | |
HRP | |
Biologically Active Proteins, Cytokine Research, Immunology | |
1437 | |
Storage of components varies. See protocol for specific instructions. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title