Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSF Antibody (SPM469), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP234774MFV500
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Human, Cynomolgus Monkey, Macaque samples. It is validated for Flow CytometrySpecifications
GM-CSF | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey, Macaque | |
Purified |
Flow Cytometry | |
SPM469 | |
Flow Cytometry | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
Recombinant human GM-CSF protein (Uniprot: P04141) | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology | |
1437 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction