Learn More
Invitrogen™ GM-CSF Monoclonal Antibody (404CE 10A8), Biotin
Mouse Monoclonal Antibody
Supplier: Invitrogen™ AHC2919

Description
This antibody can be used as a detection antibody in ELISA applications.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
Specifications
GM-CSF | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5mg/mL BSA and 0.5mg/mL sodium azide; pH 7.4 | |
P04141 | |
CSF2 | |
Purified recombinant human GM-CSF. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
404CE 10A8 | |
Biotin | |
CSF2 | |
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim | |
Mouse | |
Protein A | |
RUO | |
1437 | |
4°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.