Learn More
Invitrogen™ GM-CSF Monoclonal Antibody (MP1-31G6), Biotin, eBioscience™, Invitrogen™
Description
Description: The MP1-31G6 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF), a 14 kDa cytokine secreted by activated T cells and macrophages. Applications Reported: MP1-31G6 has been reported for use in detection of mouse GM-CSF by ELISA. Applications Tested: The MP1-31G6 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of mouse GM-CSF (Granulocyte/Macrophage-Colony Stimulating Factor, GMCSF) in combination with the purified MP1-22E9 (14-7331) antibody for capture and recombinant mouse GM-CSF (14-8331) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | GM-CSF |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MP1-31G6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | CSF2 |
| Gene Accession No. | P01587 |
| Gene Alias | colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.