Learn More
Description
Specifications
Specifications
| Concentration | LYOPH |
| Gene ID (Entrez) | 1437 |
| Purification Method | SDS-PAGE |
| Quantity | 20μg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Symbol | CSF2 |
| Product Type | GM-CSF |
| Conjugate | Unconjugated |
| Species | Human |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
