Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSF Rat, Clone: BVD2-21C11, Novus Biologicals™

Rat Monoclonal Antibody
$280.00 - $536.00
Specifications
Antigen | GM-CSF |
---|---|
Clone | BVD2-21C11 |
Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP232967F
![]() |
Novus Biologicals
NBP2329670.02MG |
0.02 mg |
Each for $280.00
|
|
|||||
NBP232967A
![]() |
Novus Biologicals
NBP2329670.1MG |
0.1 mg |
Each for $536.00
|
|
|||||
NBP232967B
![]() |
Novus Biologicals
NBP2329670.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Neutralization.Specifications
GM-CSF | |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
P04141 | |
1437 | |
Recombinant human GM-CSF protein | |
Primary | |
Store at 4C. | |
22 kDa |
BVD2-21C11 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rat | |
Cytokine Research, Immunology | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
CSF2 | |
IgG2a κ | |
Protein A purified | |
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title