Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ GM-CSF Recombinant Human Protein
Click to view available options
Quantity:
1 mg
Description
- Involved in many biological responses including growth and development of granulocyte and macrophage progenitor cells
- For stimulation and initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils
- ED50 range = 0.02 to 0.1 ng/mL, determined by dose dependent proliferation of human TF-1 cells
- Optimal concentration for individual application should be determined by dose response assay
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Molecular Weight (g/mol) | 14 kDa |
| Purification Method | Gel-Purified |
| Quantity | 1 mg |
| Source | Human |
| Gene Alias | GM-CSF |
| Product Type | Recombinant Protein |
| Species | Human |
| Recombinant | Recombinant |
| Content And Storage | Lyophilized protein can be stored at 2-8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze and thaw cycles. |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction