Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSFR alpha Rabbit anti-Human, Clone: 15, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289611100UL
Description
GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Human samples. It is validated for ELISA.Specifications
| GM-CSFR alpha | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 15 | |
| ELISA 1:5000-1:10000 | |
| CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838 | |
| Recombinant Human GM-CSFR alpha protein | |
| 100 μg | |
| Cancer | |
| 1438 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction