Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM130/GOLGA2 Antibody (2059D), Biotin, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB8199B
Description
GM130/GOLGA2 Monoclonal specifically detects GM130/GOLGA2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GM130/GOLGA2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GOLGA2 | |
Partial recombinant human GOLG-A2 protein (amino acids 528-606) [UniProt Q08379] | |
100 μL | |
Cellular Markers, Golgi Apparatus Markers | |
2801 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
2059D | |
Western Blot, Immunohistochemistry | |
130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction