Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM130/GOLGA2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP253420MFV500
Description
GM130/GOLGA2 Polyclonal antibody specifically detects GM130/GOLGA2 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
GM130/GOLGA2 | |
Polyclonal | |
Flow Cytometry | |
130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
Partial recombinant human GM130/GOLGA2 protein (amino acids 528-606). [UniPro Q08379] | |
0.1 mL | |
Cellular Markers, Golgi Apparatus Markers | |
2801 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction