Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM130/GOLGA2 Antibody (NN 2C10/1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002622AF405
Description
GM130/GOLGA2 Monoclonal specifically detects GM130/GOLGA2 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GM130/GOLGA2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GOLGA2 | |
Recombinant purified rat GM130/GOLGA2 | |
0.1 mL | |
Cellular Markers, Golgi Apparatus Markers | |
2801 | |
Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
NN 2C10/1 | |
Western Blot, Immunohistochemistry | |
130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross react with human protein. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction