Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM130/GOLGA2 Antibody (SR1125), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321627100UL
Description
GM130/GOLGA2 Monoclonal antibody specifically detects GM130/GOLGA2 in Human, Mouse, Rat, Bovine, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
GM130/GOLGA2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine, Monkey | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1125 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
A synthesized peptide derived from human GM130/GOLGA2 (Uniprot #: Q08379) | |
100 μg | |
Cellular Markers, Golgi Apparatus Markers | |
2801 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction