Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM130/GOLGA2 Antibody (NN 2C10/1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002622AF594
Description
GM130/GOLGA2 Monoclonal specifically detects GM130/GOLGA2 in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| GM130/GOLGA2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GOLGA2 | |
| Recombinant purified rat GM130/GOLGA2 | |
| 0.1 mL | |
| 2801 | |
| Rat | |
| IgG2a κ |
| Western Blot, Immunohistochemistry | |
| NN 2C10/1 | |
| Western Blot, Immunohistochemistry | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Does not cross react with human protein. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction