Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM130 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565030
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5030 Supplier Bethyl Laboratories, Inc Supplier No. A303401A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

GM130 is a putative cis-Golgi matrix protein that belongs to the golgin family of autoantigens possessing extensive coiled-coil domains. This protein interacts with GRASP65 and acts as a receptor for the vesicle docking protein p115 during vesicle fusion.

Specifications

Antigen GM130
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GOLGA2
Gene Accession No. Q08379
Gene Alias Golgin-95
Gene Symbols GOLGA2
Host Species Rabbit
Immunogen Between 350 and 400
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2801
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.