Learn More
Invitrogen™ GM130 Recombinant Rabbit Monoclonal Antibody (10H5L5)
Description
This antibody is predicted to react with Mouse, Rat, Bovine.
Specifications
Specifications
| Antigen | GM130 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 10H5L5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | Golga2 |
| Gene Accession No. | Q08379 |
| Gene Alias | 130 kDa cis-Golgi matrix protein; AW555139; cis-Golgi matrix protein GM130; Gm130; GM130 autoantigen; GOLGA2; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; golgin A2; Golgin subfamily A member 2; Golgin subfamily A member 2; LOW QUALITY PROTEIN: golgin subfamily A member 2; golgin-95; mKIAA4150; RP11-395P17.5; SY11 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.