Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gm13178 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569336
Description
This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 92%; Horse: 85%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 92%.
Gm13178 gene ontology annotations related to this gene include carboxylic ester hydrolase activity.
Specifications
Gm13178 | |
Polyclonal | |
Unconjugated | |
Gm13178 | |
Gm13178; Gm437; OTTMUSG00000010750; predicted gene 13178; uncharacterized protein LOC546849 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
546849 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
Gm13178 | |
synthetic peptide corresponding to a region of mouse Gm13178. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction