Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM2 Ganglioside Human anti-Human, Clone: L55-81, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP2812770.05MG
This item is not returnable.
View return policy
Description
GM2 Ganglioside Monoclonal antibody specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Cell depletionSpecifications
GM2 Ganglioside | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Depletion Assay | |
GM2 (ganglioside), Tay-Sachs ganglioside | |
L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma. | |
0.05 mg | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Depletion Assay | |
L55-81 | |
Unconjugated | |
PBS | |
Human | |
Affinity purified | |
RUO | |
Human | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction