Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM2 Ganglioside Human anti-Human, Clone: L55-81, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP28127725UG
Description
GM2 Ganglioside Monoclonal antibody specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Cell depletionSpecifications
GM2 Ganglioside | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Depletion Assay | |
GM2 (ganglioside), Tay-Sachs ganglioside | |
L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma. | |
25 μg | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgM κ |
ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Depletion Assay | |
L55-81 | |
Unconjugated | |
PBS | |
Human | |
Affinity purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction