Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM2 ganglioside Mouse anti-Human,Bovine, Unconjugated, Clone: GMB28, MilliporeSigma™

Catalog No. MABN25501MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25501MI 100 μL
MABN25502MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25501MI Supplier MilliporeSigma Supplier No. MABN2550100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GM2 ganglioside Monoclonal antibody specifically detects GM2 ganglioside in Human,Bovine samples. It is validated for ELISA, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM2 ganglioside
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GMB28
Conjugate Unconjugated
Gene Alias Ganglioside GM2;GM2
Host Species Mouse
Immunogen Purified GM2 ganglioside attached to Salmonella minnesota.
Purification Method Ligatrap
Quantity 100 μL
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone GMB28 is a mouse monoclonal antibody that specifically detects GM2 ganglioside.
Target Species Human, Bovine
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.