Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM2A Antibody (06), DyLight 680 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306149FR
Description
GM2A Monoclonal antibody specifically detects GM2A in Human samples. It is validated for ELISASpecifications
GM2A | |
Monoclonal | |
DyLight 680 | |
Cerebroside sulfate activator protein, ganglioside GM2 activator, GM2 ganglioside activator, GM2-AP, SAP-3GM2 ganglioside activator protein, Shingolipid activator protein 3, sphingolipid activator protein 3 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GM2A (AAA35907.1; Met1-Ile193). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
06 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
2760 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction