Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gma Dta TCR Mouse, anti-Human, R718, Clone: 11F2, BD OptiBuild™

Catalog No. BDB752023 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752023 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752023 Supplier BD Biosciences Supplier No. 752023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 11F2 monoclonal antibody specifically reacts with a framework epitope of the γ/δ TCR. The γ/δ TCR is a heterodimeric glycoprotein that is noncovalently associated with the CD3 antigen complex. The γ and δ TCR chains are composed of constant and variable regions, each encoded by distinct gene segments. The γ chain forms either disulfide-linked or non-disulfide-linked heterodimers with the δ-subunit. TCR γ/δ is present on a minor subset of T lymphocytes in peripheral blood, thymus, spleen, and lymph node. TCR γ/δ-positive T lymphocytes comprise 1% to 9% of normal peripheral blood lymphocytes and less than 2% of normal thymocytes. The 11F2 antibody is mitogenic for γ/δ-TCR-bearing T lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Gma Dta TCR
Applications Flow Cytometry
Classification Monoclonal
Clone 11F2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.