Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558845 Supplier Invitrogen™ Supplier No. PA558845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WMFLQLVPSW NGSLKQLLTE TDVWFSKQRK DFEGMAFLET EQGKPFVSVF RHLRLQYIIS DLASARIIEQ DAVVPSEWLS SVYKQQWFAM LRAEQD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

This gene encodes a nuclear envelope protein that appears to be involved in spermatogenesis, either directly or by influencing genes that play a more direct role in the process. This multi-exon locus is the homolog of the mouse and drosophila germ cell-less gene but the human genome also contains a single-exon locus on chromosome 5 that contains an open reading frame capable of encoding a highly-related protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMCL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GMCL1
Gene Accession No. Q96IK5
Gene Alias 2810049L19Rik; BTBD13; DIP; DP-interacting protein; Gcl; Gcl1; germ cell-less homolog 1; germ cell-less homolog 1 (Drosophila); germ cell-less protein; germ cell-less protein-like 1; germ cell-less, spermatogenesis associated 1; GMCL1; mGcl-1; mglc-1; SPATA29; spermatogenesis associated 29
Gene Symbols GMCL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GMCL1. Recombinant protein control fragment (Product #RP-95993).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64395
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.