Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMDS Antibody (OTI2A1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270835R
Description
GMDS Monoclonal specifically detects GMDS in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GMDS | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GMDS | |
Full length recombinant protein of human GMDS (NP_001491) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot | |
OTI2A1 | |
Western Blot | |
EC 4.2.1.47, GDP-D-mannose dehydratase, GDP-mannose 4,6-dehydratase, GMDGDP-mannose 4,6 dehydratase, SDR3E1, short chain dehydrogenase/reductase family 3E, member 1 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
2762.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction