Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMEB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560837 Supplier Invitrogen™ Supplier No. PA560837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVVLMPVSTP KPPKRPRLQR PASTTVLSPS PPVQQPQFTV ISPITITPVG QSFSMGNIPV ATLSQGSSPV TVHTLPSGPQ LFRYATVVSS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.

This gene is a member of KDWK gene family. The product of this gene associates with GMEB2 protein, and the complex is essential for parvovirus DNA replication. Study of rat homolog implicates the role of this gene in modulation of transactivation by the glucocorticoid receptor bound to glucocorticoid response elements. Two alternative spliced transcript variants encoding different isoforms exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMEB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GMEB1
Gene Accession No. Q9Y692
Gene Alias 1110050A04Rik; AI256615; AI481278; DNA-binding protein p96PIF; glucocorticoid modulatory element binding protein 1; glucocorticoid modulatory element-binding protein 1; Gmeb1; GMEB-1; P96PIF; Parvovirus initiation factor p96; PIF p96; PIF96; RGD1563208
Gene Symbols GMEB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GMEB1. Recombinant protein control fragment (Product #RP-100591).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.