Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMEB2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212763DL594
Description
GMEB2 Polyclonal antibody specifically detects GMEB2 in Human samples. It is validated for Western BlotSpecifications
GMEB2 | |
Polyclonal | |
Western Blot | |
DNA-binding protein p79PIF, glucocorticoid modulatory element binding protein 2, KIAA1269PIF p79, p79, P79PIF | |
The immunogen this antibody was made to, maps to a region between residue 480 and 530 of human Glucocorticoid Modulatory Element Binding Protein 2 using the numbering given in entry NP_036516.1 (GeneID 26205). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
26205 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction