Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GMEB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584760
Description
Immunogen sequence: ASHKCQMDRS REQYARDLAA LEQQCDEHRR RAKELKHKSQ HLSNVLMTLT PVSLPPPVKR PRLARATSGP AAMASQVLTQ SAQ.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB40, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Single nucleotide polymorphisms near ZBTB40 have been linked to differences in bone mineral density of the hip and lumbar spine, and had a significant association with low-trauma fractures suggesting that ZBTB40 may be involved in bone development and formation.
Specifications
GMEB2 | |
Polyclonal | |
Unconjugated | |
GMEB2 | |
AI839884; DNA-binding protein p79PIF; glucocorticoid modulatory element binding protein 2; glucocorticoid modulatory element-binding protein 2; GMEB2; GMEB-2; KIAA1269; P79PIF; parvovirus initiation factor p79; parvovirus initiation factor, p79; PIF p79; PIF79 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKD1 | |
GMEB2 | |
Recombinant protein corresponding to Human GMEB2. Recombinant protein control fragment (Product #RP-107577). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction