Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMEB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541540 Supplier Thermo Scientific Supplier No. PA541540
Add to Cart
Add to Cart

GMEB2 Polyclonal Antibody

GMEB2 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GMEB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GMEB2
Gene Accession No. Q9UKD1
Gene Alias AI839884; DNA-binding protein p79PIF; glucocorticoid modulatory element binding protein 2; glucocorticoid modulatory element-binding protein 2; GMEB2; GMEB-2; KIAA1269; P79PIF; parvovirus initiation factor p79; parvovirus initiation factor, p79; PIF p79; PIF79
Gene Symbols GMEB2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GMEB2 (aa 481-530).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26205
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.