Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMF-beta Antibody (44D8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259490H
This item is not returnable.
View return policy
Description
GMF-beta Monoclonal antibody specifically detects GMF-beta in Human samples. It is validated for Western Blot, ELISASpecifications
GMF-beta | |
Monoclonal | |
HRP | |
glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta | |
Recombinant human GMF-beta (1-142aa) purified from E. coli | |
0.1 mL | |
Biologically Active Proteins | |
2764 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
44D8 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction