Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMF-beta Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300104B
Description
GMF-beta Polyclonal specifically detects GMF-beta in Human samples. It is validated for Western Blot, ELISA.Specifications
GMF-beta | |
Polyclonal | |
Western Blot, ELISA | |
glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta | |
Produced in rabbits immunized with purified, recombinant GMF-beta (P60983; Met1-His142). | |
0.1 mL | |
Biologically Active Proteins | |
2764 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Biotin | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction