Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMF-beta Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300104JF646
Description
GMF-beta Polyclonal antibody specifically detects GMF-beta in Human samples. It is validated for Western Blot, ELISASpecifications
GMF-beta | |
Polyclonal | |
Western Blot, ELISA | |
glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta | |
Produced in rabbits immunized with purified, recombinant Human GMF-beta (Accession#: NP_004115.1; Met1-His142) | |
0.1 mL | |
Biologically Active Proteins | |
2764 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction