Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMF beta Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981717
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1717 Supplier Bioss Supplier No. BS11845R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein, which is phosphorylated following phorbol ester stimulation, is important for the nervous system. It causes brain cell differentiation, stimulates neural regeneration and inhibits tumor cell proliferation. Overexpression of GMF in astrocytes has been shown to enhance brain-derived neurotrohic factor (BDNF) production. GMF expression is increased by exercise, and the protein is crucial for exercise-induction of BDNF. Through BDNF production, GMF appears to play a role in neuroprotection. In thymoma, T-cell development is maintained by GMF; being produced by the tumor cells.

Specifications

Antigen GMF beta
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gmfb
Gene Accession No. P60983
Gene Alias 3110001H22Rik; 3110001O16Rik; AI851627; BOS_10924; C79176; D14Ertd630e; DNA for thyroid hormone receptor binding site (276bp); Glia maturation factor beta; glia maturation factor, beta; glia maturation factor, beta-like; GMF; Gmfb; GMF-beta; LOC100349178
Gene Symbols Gmfb
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GMF beta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2764
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.