Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMFG Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB182767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB182767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB182767 Supplier Novus Biologicals Supplier No. NBP306611F
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GMFG Polyclonal antibody specifically detects GMFG in Human samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen GMFG
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias glia maturation factor gamma, glia maturation factor, gamma, GMF-GAMMA, MGC126867
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human GMFG (O60234; Met1-Arg142).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9535
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.