Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMFG Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306611MFV610
Description
GMFG Polyclonal antibody specifically detects GMFG in Human samples. It is validated for ELISASpecifications
| GMFG | |
| Polyclonal | |
| ELISA | |
| glia maturation factor gamma, glia maturation factor, gamma, GMF-GAMMA, MGC126867 | |
| Produced in rabbits immunized with purified, recombinant Human GMFG (Accession#: O60234; Met1-Arg142) | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9535 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction