Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMFG Antibody, Novus Biologicals™
SDP

Catalog No. p-5068769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231367SS 0.025 mg
NBP231367 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231367SS Supplier Novus Biologicals Supplier No. NBP231367SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GMFG Polyclonal specifically detects GMFG in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GMFG
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3 ug/ml, Immunocytochemistry/Immunofluorescence 0.1 ug/mL
Formulation No buffer with 0.05% Sodium Azide
Gene Accession No. O60234, Q9ERL7, Q80T18
Gene Alias glia maturation factor gamma, glia maturation factor, gamma, GMF-GAMMA, MGC126867
Gene Symbols GMFG
Host Species Rabbit
Immunogen GMFG
Molecular Weight of Antigen 16.8 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9535
Test Specificity Human. Based upon the mmunogen sequence similarity, this antibody is predicted to react with Mouse (94%), Rat (91%) and Bovine (97%).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.