Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMFG Polyclonal Antibody, Invitrogen™

Catalog No. PA550345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550345 Supplier Thermo Scientific Supplier No. PA550345

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total GMFG protein.

Glia maturation factor gamma (GMFG) is a cytokine-responsive protein in erythropoietin-induced and granulocyte-colony stimulating factor-induced hematopoietic lineage development. It may be involved in glial differentiation, neural regeneration, and inhibition of tumor cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMFG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Gmfg
Gene Accession No. O60234, Q9ERL7
Gene Alias 0610039G16Rik; 2310057N07Rik; AI324845; glia maturation factor gamma; glia maturation factor, gamma; Gmfg; GMF-gamma
Gene Symbols Gmfg
Host Species Rabbit
Immunogen Full length fusion protein of Human GMFG
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63986, 9535
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.