Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545745 Supplier Invitrogen™ Supplier No. PA545745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGPDPQHHST LEQHPTATPT EIPTPQSDQR EDVAEDTKDG GGEVSSQGPE Sequence homology: Dog: 86%; Horse: 93%; Human: 100%; Rabbit: 85%.

This gene encodes a protein containing zinc finger domains. A short peptide derived from this protein is a minor histocompatibility antigen which can lead to graft rejection of male donor cells in a female recipient. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMIP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GMIP
Gene Accession No. Q9P107
Gene Alias 5031419I10Rik; ARHGAP46; GEM interacting protein; Gem-interacting protein; Gmip; RGD1565577
Gene Symbols GMIP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GMIP (aa 804-853).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51291
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.