Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559992 Supplier Invitrogen™ Supplier No. PA559992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSLLGTQSRG HFSRQPVKYP RGGVRPVTHQ LSSLALVASK LCEETPITSV PRGSLRGRGP SPAAASPEGS PLRRTPLPKH FEITQETARL LSKLD Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

This gene encodes a protein containing zinc finger domains. A short peptide derived from this protein is a minor histocompatibility antigen which can lead to graft rejection of male donor cells in a female recipient. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMIP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GMIP
Gene Accession No. Q9P107
Gene Alias 5031419I10Rik; ARHGAP46; GEM interacting protein; Gem-interacting protein; Gmip; RGD1565577
Gene Symbols GMIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GMIP. Recombinant protein control fragment (Product #RP-98902).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.