Learn More
GML Monoclonal Antibody (1E7)

Description
GML Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | GML |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1E7 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GML |
| Gene Accession No. | Q99445 |
| Gene Alias | glycosylphosphatidylinositol anchored molecule like; glycosylphosphatidylinositol anchored molecule like protein; Glycosyl-phosphatidylinositol-anchored molecule-like protein; Glycosylphosphatidylinositol-anchored molecule-like protein; GML; GPI anchored molecule like protein; LY6DL |
| Gene Symbols | GML |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.