Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GMNN Mouse anti-Human, Unconjugated, Clone: 1A8, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation H00051053M01J
Description
GMNN Monoclonal antibody specifically detects GMNN in Human samples. It is validated for IHC-P, IF, S-ELISA, ELISA, WB-Re, WB-Tr.Specifications
GMNN | |
Monoclonal | |
Unconjugated | |
GMNN | |
Gem, RP3-369A17.3 | |
GMNN (AAH05185.1, 110 a.a. ∼ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa | |
Primary | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Western Blot | |
1A8 | |
geminin, DNA replication inhibitor | |
BC005185.1 | |
Mouse | |
100 μg | |
51053 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction