Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GMNN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585964
Description
GMNN Polyclonal Antibody for Western Blot, ICC/IF, Flow
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
Specifications
GMNN | |
Polyclonal | |
Unconjugated | |
GMNN | |
AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51053, 57441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
O75496, O88513 | |
GMNN | |
Synthetic peptide within Human Geminin aa 1-50. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction