Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMNN Recombinant Rabbit Monoclonal Antibody (8H8)
GREENER_CHOICE

Catalog No. FEMA549896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549896 Supplier Invitrogen™ Supplier No. MA549896
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GMNN Recombinant Monoclonal Antibody for ICC/IF, ELISA

GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMNN
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8H8
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GMNN
Gene Accession No. O75496
Gene Alias AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3
Gene Symbols GMNN
Host Species Rabbit
Immunogen A synthesized peptide derived from human Geminin.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51053
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.