Learn More
GMP synthase Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302417AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan
In the de novo synthesis of purine nucleotides, IMP is the branch point metabolite at which point the pathway diverges to the synthesis of either guanine or adenine nucleotides. In the guanine nucleotide pathway, there are 2 enzymes involved in converting IMP to GMP, namely IMP dehydrogenase (IMPD1), which catalyzes the oxidation of IMP to XMP, and GMP synthetase, which catalyzes the amination of XMP to GMP.Specifications
GMP synthase | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P49915 | |
GMPS | |
Between 650 and 693 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GMPS | |
GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, MLL/GMPS fusion protein, glutamine amidotransferase, guanosine 5′-monophosphate synthase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8833 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.