Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMPPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543312 Supplier Invitrogen™ Supplier No. PA543312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LKAVILIGGP QKGTRFRPLS FEVPKPLFPV AGVPMIQHHI EACAQVPGMQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 100%.

GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMPPA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GMPPA
Gene Accession No. Q96IJ6
Gene Alias 1810012N01Rik; AAMR; GDP-mannose pyrophosphorylase A; Gmppa; GMPP-alpha; GTP-mannose-1-phosphate guanylyltransferase alpha; mannose-1-phosphate guanyltransferase alpha; mannose-1-phosphate guanylyltransferase (GDP)
Gene Symbols GMPPA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GMPPA (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29926
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.