Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMPPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557302 Supplier Invitrogen™ Supplier No. PA557302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKPSTFISDI INCGIYLFSP EALKPLRDVF QRNQQDGQLE DSPGLWPGAG TIRLEQDVFS ALAGQGQIYV HLTDGIWS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMPPA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GMPPA
Gene Accession No. Q96IJ6
Gene Alias 1810012N01Rik; AAMR; GDP-mannose pyrophosphorylase A; Gmppa; GMPP-alpha; GTP-mannose-1-phosphate guanylyltransferase alpha; mannose-1-phosphate guanyltransferase alpha; mannose-1-phosphate guanylyltransferase (GDP)
Gene Symbols GMPPA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GMPPA. Recombinant protein control fragment (Product #RP-102670).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.