Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMPPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595784 Supplier Invitrogen™ Supplier No. PA595784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILAVSYMSQV LEKEMKAQEQ RLGIRISMSH EEEPLGTAGP LALARDLLSE TADPFFVLNS DVICDFPFQA MVQFHRHHGQ EGSILVTKVE EPSKYGVVVC EADTGRIHRF VEKPQVFVSN KINAGMYILS PAVLRRIQLQ PTSIEKEVFP IMAKEGQLYA MELQGFWMDI GQPKDFLTGM CLFLQSLRQK QPERLCSGPG I; Positive Samples: Mouse thymus, Mouse brain; Cellular Location: Cytoplasm.

This gene is thought to encode a GDP-mannose pyrophosphorylase. The encoded protein catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, a reaction involved in the production of N-linked oligosaccharides. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMPPB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Gmppb
Gene Accession No. Q8BTZ7
Gene Alias 2.7.7.13; AI317178; cb1066; E430010H19; fc79b10; GDP-mannose pyrophosphorylase B; Gmppb; gmppl; GTP-mannose-1-phosphate guanylyltransferase beta; Mannose-1-phosphate guanyltransferase beta; MDDGA14; MDDGB14; MDDGC14; RGD1560458; Unknown (protein for MGC:134517); wu:fc79b10; zgc:92026
Gene Symbols Gmppb
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-250 of human GMPPB (NP_0688061).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 331026
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.