Learn More
Invitrogen™ GMPR Monoclonal Antibody (3G12)

Description
Peptide Sequence: MPRIDADLKL DFKDVLLRPK RSSLKSRAEV DLERTFTFRN SKQTYSGIPI IVANMDTVGT FEMAAVMSQH SMFTAIHKHY SLDDWKLFAT NHPECLQNVA VSSGSGQNDL EKMTSILEAV.
Specifications
Specifications
| Antigen | GMPR |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3G12 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with no preservative |
| Gene | GMPR |
| Gene Accession No. | P36959 |
| Gene Alias | 2310004P21Rik; AV028449; GMP reductase 1; GMPR; GMPR 1; Gmpr1; guanine monophosphate reductase; guanosine 5'-monophosphate oxidoreductase 1; guanosine monophosphate reductase; guanosine monophosphate reductase 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.